Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Polypropylene
Fournisseur: Avantor Fluid Handling
MFLX45508-30BLKEA 618 EUR InStock
MFLX45508-30BLK
Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Bulkhead, Polypropylene
Raccords et connecteurs
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from 1/16" to 1/4" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité