Masterflex® Torque Tee Sanitary Clamp Securing Devices, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX23508-03EA 365 EUR InStock
MFLX23508-03 MFLX23508-04 MFLX23508-05 MFLX23508-06 MFLX23508-07 MFLX23508-08 MFLX61126-89 MFLX38450-85
Masterflex® Torque Tee Sanitary Clamp Securing Devices, Avantor®
Sets d'outils
Ensure the security of sanitary connections with preset torque output.
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • 'T' shape creates effective leverage for fast, easy one-handed tightening
The Rubberfab Torque Tee and universal socket are designed to safely and securely install and remove sanitary clamps. Available with a family of components, the Torque Tee handle creates effective tightening leverage, even in hard-to-reach spaces. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle. The universal socket is required when using the Torque Tee.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité