Masterflex® Quick-Disconnect Fittings, Avantor®
Fournisseur: Avantor Fluid Handling
70000-37 70000-38
MFLX70000-37EA 0 EUR InStock
MFLX70000-37 MFLX70000-38 MFLX70000-27
Masterflex® Quick-Disconnect Fittings, Avantor®
Raccords et connecteurs
Disconnect and reconnect your flow path with ease.
  • Audible click lets you know the coupling is secure
  • Many hose barb sizes to choose from
  • Valved design allows you to cut off fluid flow when the fittings are disconnected
Use these fittings to quickly connect and disconnect from your flow line. Simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and the ability to be gamma sterilized.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité