Masterflex® Drum Pump Replacement Motors, Avantor®
Fournisseur: Avantor Fluid Handling
70604-03 70604-13 70604-16
MFLX70604-03EA 0 EUR InStock
MFLX70604-03 MFLX70604-13 MFLX70604-16
Masterflex® Drum Pump Replacement Motors, Avantor®
Pompe Motors
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
  • Use with Masterflex® Complete Drum and Container Pumps
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks and small storage vessels. These Masterflex® drum pump replacement motors are one component of a system that includes a pump tube and motor (order pump tubes separately). Motors are equipped with variable-speed control for added safety and control.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité