High-Efficiency Polypropylene Standard Centrifugal Pumps
Fournisseur: Avantor Fluid Handling
72009-30
MFLX72009-30EA 3390 EUR InStock
MFLX72009-30
High-Efficiency Polypropylene Standard Centrifugal Pumps
Pompe Centrifugal Pumps
Split volute casing forms a volute chamber to increase pump efficiency, reduce horsepower, and energy usage.
  • Pumps can run dry for up to one hour
  • To reduce maintenance, bearings are kept cool with heat dissipation vent holes and a large flush groove
  • Use in production processes such as filtering, spraying, washing, and etching
Pumps are set up for hard-wiring; cord and plug are not included.
Information de livraison: Includes conduit box for hardwiring.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité