Micropump® B-Mount Gear Pump Heads
Fournisseur: Avantor Fluid Handling
74004-03
MFLX74004-01EA 1780 EUR InStock
MFLX74004-01 MFLX74004-05 MFLX74004-07 MFLX74004-09 MFLX74004-11 MFLX74004-17 MFLX74004-23 MFLX74004-27 MFLX74004-29 MFLX74004-31 MFLX74004-33 MFLX74004-71 MFLX74004-73 MFLX74004-75 MFLX74004-03
Micropump® B-Mount Gear Pump Heads
Pompe Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Suction shoe designs provides higher delivery pressures and temperatures, plus increased efficiency
  • Cavity style designs with no dynamic seals for applications that cannot tolerate leaks
Magnetically driven, precision-geared pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and static seals keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of materials (due to wetted parts). Pump heads are compact and easy to service. Modular design offers flexibility—configure to your specific needs.
Call our Application Specialists for assistance.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité