Stenner Compact Economical Peristaltic Pumps
Fournisseur: Avantor Fluid Handling
74209-00 74209-02 74209-03 74209-04 74209-05 74209-06
MFLX74209-00EA 689 EUR InStock
MFLX74209-00 MFLX74209-02 MFLX74209-03 MFLX74209-04 MFLX74209-05 MFLX74209-06
Stenner Compact Economical Peristaltic Pumps
Pompe Pompes péristaltiques
Effortless tube replacement with quick-release pump head – no tools required.
  • Tubing is bonded to the fittings during manufacturing for a strong, single-piece tubing assembly
  • Run dry and self-prime without worrying about tubing or pump damage
  • Enclosed housing is wall mountable (mounting stand sold separately)
These Stenner compact economical peristaltic pumps can be run dry and are self-priming for easy pumping. Pumps are available in two models—timed and low-volume models.
Information de livraison: Includes two pump tubes, three 1/4" connecting nuts and ferrules, injection valve/injection fitting, weighted suction line strainer, 20'' (6 m) of 1/4" PE suction/discharge tubing, and AC power adapter.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité