Abaque™ Peristaltic Natural Rubber Replacement Hose
Fournisseur: Avantor Fluid Handling
75810-21
MFLX75810-32EA 339 EUR InStock
MFLX75810-32 MFLX75810-43 MFLX75810-48 MFLX75810-83 MFLX75810-21 MFLX75810-64
Abaque™ Peristaltic Natural Rubber Replacement Hose
Tuyaux Tubes pour pompe péristaltique
Highly resilient with excellent abrasion resistance and strength.
  • All listed hoses in this ad are made from Natural Rubber.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Abaque engineers / manufactures their hoses from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy, longevity, and optimal compression.
Ideal for applications where the following properties are needed:
Highly resilient, excellent abrasion resistance, the most common hose in peristaltic markets. Generally resistant with lightly corrosive chemical applications, abrasive slurries, etc.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité