Abaque™ EPDM Peristaltic Pump Replacement Hoses
Fournisseur: Avantor Fluid Handling
75810-29
MFLX75810-57EA 695 EUR InStock
MFLX75810-57 MFLX75810-29 MFLX75810-24 MFLX75810-35 MFLX75810-41 MFLX75810-46 MFLX75810-67
Abaque™ EPDM Peristaltic Pump Replacement Hoses
Tuyaux Tubes pour pompe péristaltique
High chemical resistance, especially with concentrated acids, alcohols, and ketones.
  • All listed hoses in this ad are made from EPDM.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Neptune engineers hoses the are manufactured from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy, longevity, and optimal compression.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité