Abaque™ CSM Peristaltic Pump Replacement Hoses
Fournisseur: Avantor Fluid Handling
MFLX75810-36EA 626 EUR InStock
MFLX75810-36
Abaque™ CSM Peristaltic Pump Replacement Hoses
Tuyaux Tubes pour pompe péristaltique
High resistance to oxidants, as well as to concentrated bases and acids.
  • All listed hoses in this ad are made from CSM.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Neptune engineers hoses the are manufactured from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy, longevity, and optimal compression.
Information de livraison: Includes cut length of hose, length is dependent upon the ID of the hose.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité