Masterflex® Open-Head Sensor Retrofit System, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX77097-00EA 1040 EUR InStock
MFLX77097-00
Masterflex® Open-Head Sensor Retrofit System, Avantor®
Pompe Pompes péristaltiques
Instantly shuts down the drive if the pump head is opened during operation.
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité