Masterflex® C/L® Microflex® Pump Head for Microbore Tubing, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX77123-80EA 289 EUR InStock
MFLX77123-80 MFLX77122-80 MFLX77122-82
Masterflex® C/L® Microflex® Pump Head for Microbore Tubing, Avantor®
Pompe Pump Heads Peristaltic Pump Heads
Easy tubing changes—even in tight spaces.
  • Compact design for efficient use of space
  • Microflex pump head accepts continuous lengths of tubing for clean flow paths
The Microflex® pump head features three or four rollers and a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Tube loading is quick and easy—open the head cover and release the occlusion bed in one motion—load tubing, close the cover, and you're ready to pump.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité