Masterflex® C/L® Microflex® Digital Pump System, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX77123-00-EUEA 2910 EUR InStock
MFLX77123-00-EU
Masterflex® C/L® Microflex® Digital Pump System, Avantor®
Pompe Pompes péristaltiques
Small footprint digital drive delivers accurate low flow over a wide range.
  • Digital interface with graphical LCD shows pump performance and operating mode—continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Icon driven menu allows quick, user-friendly for programming and operation
  • Dispense by Volume in ml (0.00013 to 43 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 3000 to 1 turndown ratio
  • Pump head accepts continuous lengths of microbore tubing for clean flow paths
This compact peristaltic pump features a durable, stackable ABS housing that is easy to wipe down and keep clean. The Microflex® pump head features three rollers and a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life.
Information de livraison: Includes c/L Microflex pump head and dual-voltage (115/230 VAC) benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination.
Commander maintenant
COMMANDE
SERVICES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité