Masterflex® L/S® Rigid PTFE-Tubing Pump Head, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX77390-00EA 1230 EUR InStock
MFLX77390-00
Masterflex® L/S® Rigid PTFE-Tubing Pump Head, Avantor®
Pompe Pump Heads Peristaltic Pump Heads
Ideal for high-purity fluids and aggressive organic solvents.
  • PTFE tubing is the only wetted part ensuring fluid purity and excellent chemical compatibility
  • Delivers a wide range of flow rates at pressures up to 100 psi (6.9 bar)
  • Six-roller pump head with adjustable occlusion—tighten occlusion for higher pressure, loosen for longer tubing life
The PTFE tubing pump head is ideal for applications such as filtration, chemical injection, or high-purity sampling. It is compatible with L/S drives that accept two or more pump heads with a 1/10 hp minimum. For best results operate at 1 to 300 rpm.
PTFE pump head uses 4- or 6-mm OD rigid PTFE-tubing sets—order PTFE-tubing sets separately. The 4 mm OD tubing set has a flow rate of 0.06 to 17 ml/min at 1 to 300 rpm while the 6 mm set has a flow rate of 0.25 to 65 ml/min. This pump head uses Masterflex® PTFE-tubing sets 77390-50 and 77390-60 only.
Note: do not pump gases or run dry for extended periods; run dry to prime only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité