Masterflex® L/S® Standard Digital PTFE-Tubing Pump System, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX77912-10-EUEA 3660 EUR InStock
MFLX77912-10-EU
Masterflex® L/S® Standard Digital PTFE-Tubing Pump System, Avantor®
Pompe Pompes péristaltiques
Transfer aggressive organic solvents and high-purity fluids.
  • Flow range 0.18 to 65 ml/min—low rate depends on drive rpm and 4- or 6-mm OD tubing
  • Rigid pump tubing designed to pump fluids at pressures up to 100 psi (6.9 bar)
  • Adjustable occlusion—tighten for higher pressure, loosen for longer tubing life
  • Limited chemical exposure with PTFE tubing as only wetted part
This complete Masterflex system is ideal for filtration, chemical injection, and other high pressure applications. The PTFE pump head accepts either 4- or 6-mm OD rigid PTFE-tubing sets. Note: Use Masterflex® PTFE-tubing sets only.
Information de livraison: Includes l/S® PTFE-tubing pump head 77390-00, PTFE-tubing set 77390-60, and L/S® variable-speed drive 07528-20 (3 to 300 rpm). System also includes two 1/4" NPT(M) pipe adapters.
Commander maintenant
COMMANDE
SERVICES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité