Masterflex® L/S® PTFE-Diaphragm Pump System, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX77915-10-EUEA 4010 EUR InStock
MFLX77915-10-EU
Masterflex® L/S® PTFE-Diaphragm Pump System, Avantor®
Pompe Pompes à membrane
Ideal for chemical feed/metering and applications requiring high-purity, high-pressure or both.
  • Flow range 80 to 800 ml/min—flow rate depends on drive rpm and tubing size
  • PTFE wetted parts help maintain fluid purity
  • Pump through filters or into pressurized system
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
  • Unique geometry minimizes fluid churning and dead volume for high metering accuracy without reagent contamination
  • Accurate and repeatable settings with 3-digit LED display of rpm
  • Maximize processes in a minimal footprint by stacking splash-resistant, IP33-rated drives
  • Two ways to control speed, direction, and start/stop—use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Information de livraison: Includes pTFE-diaphragm pump head 07090-42, 12 ft (3.6-m) of PTFE tubing (1/4" ID x 3/8" OD), L/S drive 07528-10, two PFA pipe adapters (1/4" NPT(F) x 3/8" OD), foot valve, mounting tools, and coupling hardware.
Commander maintenant
COMMANDE
SERVICES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité