Masterflex® L/S® Variable-Speed Air-Powered Pump Systems, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX77931-10EA 3000 EUR InStock
MFLX77931-10
Masterflex® L/S® Variable-Speed Air-Powered Pump Systems, Avantor®
Pompe Pompes péristaltiques
Use where electrical power is unsafe or unavailable.
  • Flow range 3.6 to 3400 ml/min—low rate depends on drive rpm, tubing head, and tubing size
  • Easily vary speed by adjusting air pressure with regulator
  • Rugged 3/4-hp air motor requires little maintenance and generates less heat than an electric motor
  • IP44 rated—painted steel housing protects from gentle spray
Use this variable-speed air-powered system whenever electrical power is unsafe or impractical. Careful grounding to protect from static electricity is vital for safe operation. Drive runs on 3 to 25 cfm air input at 20 to 100 psi.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité