Masterflex® Transfer Tubing, PVDF, Avantor®
Fournisseur: Avantor Fluid Handling
95100-04
MFLX95100-00EA 149 EUR InStock
MFLX95100-00 MFLX95100-02 MFLX95100-06 MFLX95100-10 MFLX95100-12 MFLX95100-14 MFLX95100-16 MFLX95100-04
Masterflex® Transfer Tubing, PVDF, Avantor®
Tuyaux Transfer Tubing
Fluoropolymer tubing at less cost.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for high-purity applications and corrosive chemical transfer—an inexpensive alternative to fluoropolymer tubing. The polyvinylidene fluoride (PVDF) material is kink resistant, has excellent chemical resistance, and is odorless and nontoxic. The resin is FDA-compliant (21 CFR 177.2600); meets USP 23 Class VI and USDA requirements. It has a temperature range of -40 to 260 °F (-40 to 127 °C). Can be
sterilized with autoclave and gamma irradiation.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité