Masterflex® Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Platinum-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
95603-24 95603-38
MFLX95603-24EA 160 EUR InStock
MFLX95603-24 MFLX95603-26 MFLX95603-28 MFLX95603-38 MFLX95603-44 MFLX95603-48 MFLX95603-40 MFLX95603-42 MFLX95603-22 MFLX95603-30 MFLX95603-34 MFLX95603-36 MFLX95603-32
Masterflex® Ismatec® Pump Tubing, 3-Stop Color-Coded Autoanalysis/Microbore, Platinum-Cured Silicone, Avantor®
Tuyaux Tubes pour pompe péristaltique
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • No leachable additives or plasticizers
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The platinum-cured silicone tubing features an ultra-smooth inner bore, excellent biocompatibility for sensitive applications, non-cytotoxic, non-pyrogenic, and non-hemolytic, low protein binding, heat-sealable, bondable, sterile-weldable, and custom moldable for assemblies. It is ideal for pharmaceutical, biotech, and general-purpose lab applications, including sterile filling and processing, cell harvest and media process systems, filtration, fermentation, cell and tissue culture transport. It features a clear appearance, has a temperature range of –66 to 135 °C, and has a pumping life of approximately 1000 hours at 100 rpm.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité