Masterflex® Transfer Tubing, Norprene® Pressure, Avantor®
Fournisseur: Avantor Fluid Handling
95715-00
MFLX95715-00EA 473 EUR InStock
MFLX95715-00
Masterflex® Transfer Tubing, Norprene® Pressure, Avantor®
Tuyaux Transfer Tubing
Flexibility at extreme temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Ideal for chemical transfer, pesticide/fertilizer delivery, water feed lines, food/dairy processing, and water purification lines. The tube features excellent resistance to acidic and alkali substances, ozone, UV light, and abrasive media. It is nonconductive, heat-sealable, non-aging, and nonoxidizing. In addition, the tube maintains flexibility even at extreme temperatures. Temperature range is -60 to 275 °F (-51 to 135 °C). Can be sterilized with autoclave.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité