Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, PharMed® BPT, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX95723-10EA 145 EUR InStock
MFLX95723-10 MFLX95723-12 MFLX95723-14 MFLX95723-18 MFLX95723-22 MFLX95723-24 MFLX95723-26 MFLX95723-28 MFLX95723-30 MFLX95723-32 MFLX95723-34 MFLX95723-36 MFLX95723-38 MFLX95723-40 MFLX95723-42 MFLX95723-44 MFLX95723-46 MFLX95723-48
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, PharMed® BPT, Avantor®
Tuyaux Tubes pour pompe péristaltique
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order two-stop color-coded and extension tubing. Tubing comes in various formulations for almost any pumping application.
The stops on the two-stopMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité