Masterflex® Transfer Tubing, Polyimide, Avantor®
Fournisseur: Avantor Fluid Handling
95820-00 95820-01 95820-02 95820-03 95820-04 95820-05 95820-06 95820-07 95820-08 95820-09 95820-10 95820-11 95820-12 95820-13
MFLX95820-00EA 74.6 EUR InStock
MFLX95820-00 MFLX95820-01 MFLX95820-02 MFLX95820-03 MFLX95820-04 MFLX95820-05 MFLX95820-06 MFLX95820-07 MFLX95820-08 MFLX95820-09 MFLX95820-10 MFLX95820-11 MFLX95820-12 MFLX95820-13
Masterflex® Transfer Tubing, Polyimide, Avantor®
Tuyaux Transfer Tubing
Provides excellent temperature stability.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
The tubing is ideal for use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers. It offers excellent chemical, heat, radiation, and abrasion resistance and high dielectric strength. Resin meets USP Class VI requirements and NEMA standards. The tube has a temperature range of –418 to 896 °F (–250 to 480 °C). Can be sterilized with autoclave, ethylene oxide or gamma irradiation.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité