Sani-Tech® Ultra Platinum-Cured Silicone Lab Tubing
Fournisseur: Avantor Fluid Handling
96201-63
MFLX96201-19EA 61.5 EUR InStock
MFLX96201-19 MFLX96201-50 MFLX96201-55 MFLX96201-60 MFLX96201-63
Sani-Tech® Ultra Platinum-Cured Silicone Lab Tubing
Tuyaux Transfer Tubing
Don't let extractables ruin your formulations.
  • Exceptionally low TOCs minimize fluid contamination
  • Ultra-pure silicone will not impart flavors or odors
  • Superior pumping life reduces downtime for tubing changes
Applications: Sterile fill and transfer, cell harvest, media processing, sanitary peristaltic pumping, production filtration and fermentation, bioreactor process lines, and pharmaceutical delivery.
Characteristics: Excellent flexibility, long pump life, non-yellowing, tasteless and odorless, nontoxic, translucent.
Certifications: Elastomer is FDA-compliant and meets USP Class VI, EP 3.1.9, and ISO 10993 biocompatibility criteria. Tubing is manufactured and packaged in a certified cleanroom.
Temperature range: -80 to 500 °F (-62 to 260 °C).
Sterilize: Autoclave, ethylene oxide, or gamma irradiation (up to 5.0 Mrad).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité