Masterflex® B/T® PerfectPosition® Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX96423-92EA 6930 EUR InStock
MFLX96423-92
Masterflex® B/T® PerfectPosition® Pump Tubing, BioPharm Platinum-Cured Silicone, Avantor®
Tuyaux Tubes pour pompe péristaltique
Ensure optimal performance from your Masterflex® B/T series pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Platinum-cured silicone
  • Ultra-smooth inner surface
  • Fewer extractables
  • Minimizes particle entrapment
  • Perfect for lab, biotechnology, & pharmaceutical applications
  • Long tubing life
  • USP Class VI; Exceeds 3A Standards
  • FDA Compliant
Formulation: Tygon 3350 silicone, platinum cured.
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications.
Characteristics: Ultra-smooth; minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, and nontoxic. Transparent.
NOTE:The below tubing is not designed to work with the B/T Perfect Position Pump Systems.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité