Masterflex® B/T® PerfectPosition® Pump Tubing, BioPharm Plus Platinum-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX96445-87EA 968 EUR InStock
MFLX96445-87
Masterflex® B/T® PerfectPosition® Pump Tubing, BioPharm Plus Platinum-Cured Silicone, Avantor®
Tuyaux Tubes pour pompe péristaltique
Ensure optimum performance from your Masterflex® pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Optically inspected, factory tested
  • Ensures flow rates are accurate and promotes longer service life
  • Seven different tubing formulations—choose tubing based on chemical compatibility, life, and pressure or vacuum needs
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
Change Notice CCN-2023-173 covers all products shipped after Jan 1st 2024.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité