Masterflex® Transfer Tubing Spool, PharMed® BPT, Avantor®
Fournisseur: Avantor Fluid Handling
MFLX96900-06EA 2990 EUR InStock
MFLX96900-06
Masterflex® Transfer Tubing Spool, PharMed® BPT, Avantor®
Tuyaux Transfer Tubing
Protect your light-sensitive samples.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 to 165 °F (-46 to 73 °C). Can be sterilized with ethylene oxide or autoclave.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité