Masterflex® Transfer Tubing Spool, Platinum-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
98906-12
MFLX98906-00EA 9310 EUR InStock
MFLX98906-00 MFLX98906-02 MFLX98906-04 MFLX98906-06 MFLX98906-08 MFLX98906-10 MFLX98906-12
Masterflex® Transfer Tubing Spool, Platinum-Cured Silicone, Avantor®
Tuyaux Transfer Tubing
Low compression formulation keeps its shape and cleanliness.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Reduce unpacking time, save money on shipping and ensure that you always have enough tubing on hand with bulk tubing. Platinum-cured silicone spools are sold in 300 ft or 500 ft spools.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité