HPLC precolumn filters
Fournisseur: MICROSOLV TECH CORP
49650-005
MICS49650-005EA 228 EUR InStock
MICS49650-005
HPLC precolumn filters
Filtres Filtres de précolonnes
The flter protects columns from particulates and extends the column life-time.
During use, as column back-pressure increases to unacceptable levels quickly remove the disposable precolumn filter. This will minimise instrument downtime and extend the life of your column.
The precolumn body contains a stainless steel filter element and does not introduce any extra column volume so that the chromatography is not affected.
With its convenient direct connect design, changeover time is measured in seconds and no tools or wrenches are needed.
Fittings compatibility: 1/16th 10-32
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité