HPLC columns, Cogent Diamond Hydride™
Fournisseur: MICROSOLV
70000-05P-2 70000-15P 70200-05P-2 70000-7.5P 70000-P30-100 70200-10P-2 70000-HG9 70000-05P 70000-05P-1 70000-10P 70000-10P-1 70000-10P-2 70000-15P-1 70000-15P-2 70000-25P 70000-HG5 70000-HG6 70000-P21-100 70000-P21-150 70000-P21-250 70000-P21-50 70000-P30-250 70000-P30-50 70000-SP100 70000-SP150 70000-SP250 70000-SP50 70200-10P-1 70200-15P-1 70200-15P-2 70200-HG3
MICS70000-05P-2EA 933 EUR InStock
MICS70000-05P-2 MICS70000-15P MICS70200-05P-2 MICS70000-7.5P MICS70000-P30-100 MICS70200-10P-2 MICS70000-HG9 MICS70000-05P MICS70000-05P-1 MICS70000-10P MICS70000-10P-1 MICS70000-10P-2 MICS70000-15P-1 MICS70000-15P-2 MICS70000-25P MICS70000-HG5 MICS70000-HG6 MICS70000-P21-100 MICS70000-P21-150 MICS70000-P21-250 MICS70000-P21-50 MICS70000-P30-250 MICS70000-P30-50 MICS70000-SP100 MICS70000-SP150 MICS70000-SP250 MICS70000-SP50 MICS70200-10P-1 MICS70200-15P-1 MICS70200-15P-2 MICS70200-HG3
HPLC columns, Cogent Diamond Hydride™
Colonnes pour chromatographie
These useful HPLC columns have a silica hydride surface similar to other TYPE-C™ phases but the uniqueness of Diamond Hydride™ is that they have a very small amount of carbon on the surface that adjusts the hydrophobicity to a small but important level. This is an excellent choice for small, polar compounds by LCMS or HPLC.
  • No water shell
  • Fast equilibration in HILIC mode
  • Excellent for polar compounds and some non-polar compounds
Popular choice of scientists working in metabolomics or bioanalytical with LCMS for compounds such as amino acids, organic acids, carbohydrates and very polar small molecules. Precise methods are easily developed even with complex sample matrices including biological fluids such as plasma, urine, saliva and other bio matrices. This is an excellent choice to replace HILIC methods when precision and speed are desired. Lack of water in these columns gives them the ability to equilibrate after only three column volumes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité