Detergents for glassware washers, ProCare Lab 11 AP
Fournisseur: MIELE
10266540
MIEL10266540EA 90 EUR InStock
MIEL10266540
Detergents for glassware washers, ProCare Lab 11 AP
Produits nettoyants Détergents et Nettoyants de Surfaces
Powder, alkaline detergent for machine based cleaning of laboratory glassware and laboratory utensils.
  • Removes inorganic and organic soils
  • Free from phosphates
  • Can be used in the water hardness range up to 6°dH
ProCare Lab 11 AP is suitable for general purpose use in water laboratories, serological institutes and in laboratories in the phosphate industry.
Note: Suitable for stainless steel, laboratory glassware, plastics such as PE, PP, PVDF, PTFE. Not suitable for use on non ferrous metals, light alloys, anodised aluminium, and aluminium.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité