EZ-Pak® Membrane Dispenser Curve
Fournisseur: Merck
EZCURVE01
MILFEZCURVE01EA 4150 EUR InStock
MILFEZCURVE01
EZ-Pak® Membrane Dispenser Curve
Membrane Dispensers
EZ-Pak® Membrane dispenser curve has robust components selected for durability and high-throughput dispensing. New design with cover allows you to load a cartridge in less than 30 seconds. Membrane is consistently well-positioned for easy transfer with forceps.
  • High-speed sterile membrane dispensing
  • No-touch operation
  • One-handed membrane transfer
  • The smooth shape facilitates cleaning
  • The plastic material is compatible with a large range of disinfection agents
  • Lightweight and portable
No need to press a lever, the durable motor will dispense one membrane in less than one second. Move your hand toward the sensor - no touch necessary and a membrane is unwrapped, dispensed and automatically positioned for transfer with forceps. Software controls prevent the accidental dispensing of multiple membranes.
The dispenser can work with or without connection to a power outlet. When the Lithium ion battery
is fully charged, more than 10000 membranes can be dispensed. After 10 minutes idle time, a standby mode is activated to save energy.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité