Samplicity® Filtration Systems
Fournisseur: Merck
SAMPSYSGR
MILFSAMPSYSGREA 1430 EUR InStock
MILFSAMPSYSGR
Samplicity® Filtration Systems
Filtres Unités de filtration sous vide
Vacuum filtration system for sample preparation for HPLC. The system’s Millex Samplicity® filters have a unique funnel shape for easy pipette loading and are provided in strips of four for faster loading. Filter strips are perforated for use with fewer samples.
  • Non-sterile
  • Ergonomic benefits
  • Efficient filtration of hard-to-filter samples
  • High recovery of small volume samples
  • Minimal evaporation of solvent
Material: polyester component, polypropylene, silicone component.
Application: HPLC and analytical sample preparation.
The Samplicity® Filtration System is ideal for QC and R&D labs, along with other analysts studying:
Drug dissolution - mandatory testing of the dissolution rate of solid dosage forms using an in vitro test
Food safety - testing foods and beverages for unknown and known toxins, including glycol, melamine, and cyanobacteria
Cosmetics - separation and detection of cosmetic ingredients and formulations
Biofuels - analysis and extraction of lipids from algae and other biomass
Pharmacokinetic/pharmacodynamic (PK/PD) - quantification of interactions of drugs with the body with respect to time
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité