Stericap™ PLUS Universal Bottle-Top Filter
Fournisseur: Merck
SCGPCAPRE
MILFSCGPCAPREEA 254 EUR InStock
MILFSCGPCAPRE
Stericap™ PLUS Universal Bottle-Top Filter
Filtres Unités de filtration sous vide
The Stericap™ PLUS Universal bottle-top filter device with Millipore Express® PLUS membrane provides fast vacuum filtration and low protein binding without the need for a prefilter in most cases. It filters quickly, without clogging, even viscous solutions typical of serum-containing media. This non-cytotoxic, non-pyrogenic device can be used to fill any vacuum-rated bottle or other container with an inner neck diameter of 20 to 67 mm. This allows maximum versatility, capacity, speed, and flowthrough efficiency when you need to filter larger volumes into containers with openings of various sizes.
  • Material: Styrene/Polypropylene/Polyethylene
  • Vented to help prevent filter air lock
  • Features fast-flowing, low protein binding Millipore Express® PLUS membrane
  • Ideal for fast sterilisation of tissue culture media, serum, buffers, or other biological solutions
  • High volume capacity for larger and less frequent preparation steps
  • Easy flow control button so the filter can be easily transferred from bottle to bottle without turning off the vacuum each time
  • Free of cytotoxic and pyrogenic contaminants for robust cell culture and sensitive downstream applications
Applications: Sterilisation of aqueous solutions, media preparation, buffer preparation.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité