Steritop® E-GP Funnel-less Sterile Vacuum Filtration System
Fournisseur: Merck
SEGPT0038 SEGPT0045
MILFSEGPT0038EA 170 EUR InStock
MILFSEGPT0038 MILFSEGPT0045
Steritop® E-GP Funnel-less Sterile Vacuum Filtration System
Filtres Unités de filtration sous vide
Work with ease and filter with confidence with Stericup® and Steritop® filtration systems. The Stericup® E and Steritop® E sterile filters are designed as a greener alternative for vacuum sterile filtration that significantly reduces plastic and packaging.
  • The filter collar disconnects from the receiver bottle with just a quarter turn reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood
  • Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité