Durapore® PVDF white membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.
- Can be sterilised by ethylene oxide and gamma irradiation
- Available in several pore sizes (both hydrophilic and hydrophobic)
- Have very low protein binding to minimize interaction with your sample and maximise recovery
Filter surface: plain
Bacterial endotoxins: 0,5 EU/ml
Gravimetric extractables: <0,5%