Milli-Q® CLX 7040 CLRW Water Purification System
Fournisseur: Merck
ZAFS51040
MILLZAFS51040EA 30260 EUR InStock
MILLZAFS51040
Milli-Q® CLX 7040 CLRW Water Purification System
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
Milli-Q® CLX 7000 analyser feed systems produce up to 3000 L/day of pure water from tap.
  • Consistent CLRW (Clinical laboratory reagent water) quality water that exceeds CLSI® standards
  • Advanced and sustainable water purification technologies, including the Elix® EDI module and water-saving E.R.A.® technology
  • User-friendly touchscreen
  • Quiet, compact and mobile
  • Digital services for remote monitoring and traceability
  • Store in dry location
Milli-Q® Connect remote services simplifies system management, supports full data traceability and access, and facilitates audit preparation and lab accreditation.
Water filtered through 0,22 μm sterilising-grade membranes and exposed to bactericidal UV lamp.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité