Milli-Q® SQ 200 Water Purification Systems
Fournisseur: Merck
ZSQ200U0T0
MILLZSQ200U0T0EA 3380 EUR InStock
MILLZSQ200U0T0
Milli-Q® SQ 200 Water Purification Systems
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
The Milli-Q® SQ 200 water purification system delivers ultrapure (Type 1) water from the SQ Switch 3,5 L tank filled with pure reverse osmosis (RO) water. It benefits from a flexible handset dispenser to deliver ultrapure water. This dispensing module must be associated with a Milli-Q® SQ 2Series family pure RO production station to refill its mobile SQ Switch tank.
  • Ultrapure (Type 1) water dispensing module from one 3,5 L Switch tank
  • Flexible dispensing
  • For elementary lab use
  • Compact - mobile - scalable
  • Simply plug in, use and maintain
Simple, manual handset dispenser allows any new or untrained user to deliver ultrapure water at variable flow rate, from up to 1,6 L/min to drop by drop. Reliable 18,2 MΩ·cm resistivity ultrapure (Type 1) water quality is indicated at a glance by a coloured LED, and TOC is reduced by an integrated photo-oxidation UV lamp
This Milli-Q® SQ 200 dispensing module allows to set or scale up 5 of 6 preset configurations available for the Milli-Q® SQ 2Series family: Milli-Q® SQ 240, Milli-Q® SQ 240L, Milli-Q® SQ 240XL, Milli-Q® SQ 240C, and Milli-Q® SQ 240CV.
Fast (as little as 6 min) refilling of the mobile SQ Switch tank with freshly produced pure RO water thanks to the high-flow RO cartridge (up to 42 L/h) of the pure water production station; RO water recovery loop saves 50% of the rejected water; automatic RO membrane rinsing maintains its high performance.
Compact set up and self-guided installation manual and videos let you plug the system wherever is convenient for your lab activity, and move it later if needed by yourself.
Unlimited scalability with one pure RO water production station able to feed multiple autonomous Milli-Q® SQ 200 ultrapure dispensing modules (just plug in an electrical outlet); Supports lab activity expansion while limiting budget investment. Individual Twist and Lock cartridge replacement, for each exhausted purification step, simplifies maintenance and limits running cost. Compact size plus water and energy savings support lab sustainability efforts.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité