Millicell® Cell Culture Inserts, Hanging, PET Membrane
Fournisseur: Merck
PIRP30R48 PIRP12R48
MILXPIRP30R48EA 392 EUR InStock
MILXPIRP30R48 MILXPIRP12R48
Millicell® Cell Culture Inserts, Hanging, PET Membrane
Microplaques Inserts pour plaques de cultures
With Millicell inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell inserts make it possible to study both sides of the cell monolayer.
  • Hanging inserts ideal for co-culturing and permeability assays
The inserts are easily prepared for SEM and TEM visualising techniques, and they are compatible with cellular and/or fluorescent stains.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité