QBT™ Fatty acid uptake assay kit
Fournisseur: Molecular Devices
R8132 R8133 R6132
MLDVR8132EA 1210 EUR InStock
MLDVR8132 MLDVR8133 MLDVR6132
QBT™ Fatty acid uptake assay kit
Tests Tests cellulaires
QBT™ Fatty Acid Uptake Assay Kit is a homogeneous assay amenable to high-throughput screening. The kits deliver pre-optimized, fluorescence-based formulations to expedite assay development and screening of fatty acid transporters.
  • One-step, homogeneous assay format enables high-throughput screening
  • Fluorescent assay with proprietary quench technology allows users to monitor real-time uptake kinetics in live cells
  • Replaces cumbersome and time-consuming fluorescent and radioactive assay protocols
  • Elimination of radioactive compounds reduces disposal costs and eliminates safety risks associated with radioactive assays
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité