Cell integrity kit, EarlyTox™
Fournisseur: Molecular Devices
R8216 R8217 R8213 R8214 R8215
MLDVR8216EA 226 EUR InStock
MLDVR8216 MLDVR8217 MLDVR8213 MLDVR8214 MLDVR8215
Cell integrity kit, EarlyTox™
Tests Tests cellulaires
The EarlyTox™ Cell Integrity Kit enables the differentiation of live cells from dead cells via fluorescent labeling. This is useful for the rapid quantification of cell viability when used with the SpectraMax® MiniMax™ 300 Imaging Cytometer or other fluorescence-based cellular imaging instruments.
  • Useful for the rapid quantification of cell viability
  • Higher signal allows shorter exposure times for fast results
  • Designed to work with many cell types - both adherent and non adherent
  • Streamlined image acquisition and analysis on a single system
The EarlyTox® Cell Integrity Kit is an optimised set of reagents that simplifies the identification of live and dead cells. It can be used to measure the effects of different treatments on cell viability and to evaluate toxic effects mediated through a variety of mechanisms, including apoptosis and necrosis. The kit is designed to work with many cell types, both adherent and non-adherent. The protocol's simple workflow and reagent performance make it amenable to high-throughput screening.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité