Scotch-Weld™ DP190 Epoxy Adhesive Grey
Fournisseur: 3M
190.50
MMMA190.50EA 487 EUR InStock
MMMA190.50
Scotch-Weld™ DP190 Epoxy Adhesive Grey
Adhésifs
Scotch-Weld™ Epoxy adhesive DP190 is a two-part epoxy adhesive that delivers exceptional performance with high shear and peel strength. Extremely compatible, it is formulated to bond to a wide range of materials, including metals, ceramics, wood, fibreboard, glass, rubber, and many plastics.
  • Apply with ease with a low viscosity formulation
  • Two-part epoxy adhesive is highly compatible with many different materials
  • Formulated with low viscosity for easy dispensing
Scotch-Weld™ Epoxy Adhesive DP190 adheres with tough, strong bonds and offers an extended work life. The adhesive is designed with a low viscosity formulation to give additional time for adjustment and make application easy.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité