3M™ Professional Masking Tape
Fournisseur: 3M
209036SW
MMMA209036SWEA 6.7 EUR InStock
MMMA209036SW
3M™ Professional Masking Tape
Rubans adhésifs
3M™ Professional Masking Tape 2090 is a universal tape that can be used to mask various surfaces such as glazing, glass, wood, plastic, tiles and metal.
  • UV and water resistant
  • Ideal for glass, wood, plastic, tiles and metal, indoors and outdoors
  • The masking tape is made from PEFC certified paper, whose forests are managed sustainably and responsibly
Use 3M™ Professional Masking Tape 2090 to achieve clean paint lines on all interior and exterior surfaces. It adheres evenly to most surfaces without bleed-through, ensuring perfect paint lines. It can be removed cleanly without residue for up to 14 days after application.
Adhesive type: Acrylic
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité