Organic Vapour Monitor, 3500+
Fournisseur: 3M
3500PLUS
MMMA3500PLUSEA 346 EUR InStock
MMMA3500PLUS
Organic Vapour Monitor, 3500+
Systèmes de détection de gaz
3M™ Organic vapour monitor 3500+ is a cost-effective and convenient way to perform personal air sampling for many organic vapours. Simply clip the device to a shirt lapel, collar, or pocket. No batteries, hoses, or pumps or needed to use the monitor to sample air.
  • State of the art monitor design
  • Simple, cost-effective, and convenient
  • High sample capacity for sampling a broad range of organic vapours
  • Monitors are small and lightweight, and will not interfere with employee activities
  • Documentation for industrial hygienists and IH labs available upon request
  • Analysis is not included
Knowing the specific contaminant and exposure levels is important to determine the appropriate respirator and filter for any work environment. This information will also help in estimating the life of selected 3M™ filters.
The organic vapour monitor 3500+ uses diffusion, rather than a pump, to collect an air sample with an accuracy of +/-25%, at a 95% confidence level for many workplace organic vapours, for both personal or area sampling. It contains a single charcoal sorbent wafer for easy collection of organic vapours and can be used for most applications. After following all instructions, send the monitor to be analysed in the lab of your choice. This monitor has a 2-year shelf life from its production date.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité