Organic Vapour Monitor, 3501+
Fournisseur: 3M
3501PLUS
MMMA3501PLUSEA 214 EUR InStock
MMMA3501PLUS
Organic Vapour Monitor, 3501+
Systèmes de détection de gaz
3M™ Organic vapour high sampling rate monitor 3501+ is a cost-effective and convenient way to perform personal air sampling for low levels of organic vapours and short-term exposure limit (STEL) sampling. Simply clip the device to a shirt lapel, collar, or pocket. No batteries, hoses, or pumps or needed to use the monitor to sample air.
  • State of the art monitor design
  • Simple, cost-effective and convenient
  • Highest diffusive sampling rate available for monitoring low contaminant levels and STEL sampling
  • Monitors are small and lightweight and will not interfere with employee activities
  • Documentation for industrial hygienists and IH labs available upon request
  • Analysis is not included
Knowing the specific contaminant and exposure levels is important to determine the appropriate respirator and filter for any work environment. This information will also help in estimating the life of selected 3M™ filters.
The Organic Vapour High Sampling Rate Monitor 3501+ uses diffusion, rather than a pump, to collect an air sample with an accuracy of ±25%, at a 95% confidence level for many workplaces organic vapours, for both personal or area sampling. It contains 562 sampling ports to generate high sampling rates and is designed to detect organic vapours at low concentrations or during short-term (STEL) sampling when low detection limits are important. Because of the high sampling rate, this monitor may exceed capacity at high concentrations or during long sampling times.
After following all instructions, send the monitor to be analysed in the lab of your choice. This monitor has a 2-year shelf life from its production date.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité