Bande de signalisation de danger 767
Fournisseur: 3M
767IRW50IW
MMMA767IRW50IWEA 157 EUR InStock
MMMA767IRW50IW
Bande de signalisation de danger 767
Rubans adhésifs
3M™ Hazard Warning Tape 767 is a red and white striped vinyl tape. This tape is used for marking hazardous areas, protruding equipment, or preventing accidents in factories, warehouses, and hospitals.
  • Use for taping in all non-critical applications
  • Conformable backing makes vinyl tape ideal for use on curved and irregular surfaces
  • Rubber adhesive provides good instant adhesion on many surfaces
  • Maintains vivid colour over time for long-lasting quality
  • Can be applied by hand or with floor applicator
  • Ideal for marking applications in light traffic areas
Identify and warn with the distinctive Hazard Warning Tape 767. This red and white striped vinyl tape is coated with a rubber adhesive and is designed for use as a marking tape for a variety of warning applications. The rubber adhesive offers quick stick and good holding strength on many substrates. Unlike messy paint solutions, this vinyl tape is quick to install and easy to clean up. Conformability and instant adhesion makes this product a good choice for applications on floors, equipment, walls, and pipes.
Recommended applications include, floor marking, marking equipment pinch points, temporary floor marking around spills, and safety markings.
This tape features abrasion resistance to excel in light-duty identification of equipment and parts. These features, combined with easy removal properties, often make it a faster, more versatile, and less costly option than painting when used in marking and coding applications.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité