Ruban polyester 8992
Fournisseur: 3M
8992
MMMA8992EA 37.8 EUR InStock
MMMA8992
Ruban polyester 8992
Rubans adhésifs
3M™ Polyester Tape 8992 is a general-purpose tape that excels in high temperature masking operations such as powder coat painting. Coated with a silicone adhesive, this tape bonds to a variety of difficult surfaces. It is chemical-resistant and performs under a wide range of temperatures and environments.
  • Works in demanding conditions and high temperatures
  • Polyester film offers excellent thermal and chemical resistance while maintaining flexibility
  • Thicker version of 3M™ Polyester Tape 8991
  • Silicone adhesive provides clean removal from a variety of surfaces
  • Translucent green backing allows easy identification and positioning
Coated with a silicone adhesive, this tape functions well in masking for powder coating to provide sharp paint lines. It removes cleanly and in one piece from most surfaces, avoiding slivering and breaking. This tape is a thicker version of 3M™ Polyester Tape 8991. With excellent chemical resistance, this tape works effectively in chemical processes and demanding work conditions, such as anodizing. The pressure sensitive silicone adhesive performs under a broad range of temperatures, ranging from −60 to +400 °F (−50 to +204 °C).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité