Rubans, double face, 9191
Fournisseur: 3M
9191
MMMA9191EA 691 EUR InStock
MMMA9191
Rubans, double face, 9191
Rubans adhésifs
3M™ double coated tape 9191 is a general-purpose double-coated tape. It features a high tack, high initial adhesion rubber pressure sensitive adhesive, a non-woven carrier, and an easy to remove siliconised paper liner.
  • General-purpose, double sided tissue tape
  • High tack, high initial adhesion
  • Pressure sensitive adhesive
  • Carpet tape
  • Cloth back material
With excellent balance between high tack/initial adhesion and good room temperature shear resistance as well as good removability, 9191 tape has been developed as a carpet tape. It is, however, necessary to systematically evaluate surface compatibility before starting an application.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité