Tape, Double Coated, 9731
Fournisseur: 3M
9731
MMMA9731EA 6460 EUR InStock
MMMA9731
Tape, Double Coated, 9731
Rubans adhésifs
The tape is developed with a high-performance acrylic adhesive on the face side and a silicone adhesive on the liner side of a 1 mil clear PET carrier.
  • Double linered - Adhesive 350
  • Ideal for attaching silicone rubber elastomers and foam
Features acrylic and silicone adhesives for specialised applications such as silicone gasketing. A thin polyester carrier provides dimensional stability and improved handling with ease of die cutting and lamination compared to adhesive transfer tapes.
It is excellent for demanding applications where silicone materials must be bonded to a range of substrates.
Core size: 76 mm
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité