Breathing Tube Adapters
Fournisseur: 3M
ADP-03
MMMAADP-03EA 227 EUR InStock
MMMAADP-03
Breathing Tube Adapters
Respirators Systèmes de respiration
The QRS breathing tube adapter ADP-03 is designed to allow the physical connection between certain Versaflo™ headtops with the QRS (Quick Release System) fitting to the 32 mm thread fitting found on specific 3M™ powered air respirator system, PF-600 series breathing tubes.
  • Innovative QRS (Quick Release Swivel) connects quickly and helps avoid kinks in the breathing tube
Create reliable connections between the powered air turbo and headtop when using a 32 mm threaded breathing tube with 3M™ QRS breathing tube adapter, ADP-03. This adapter can be used with 3M™ powered air respirator system PF-600 series alongside certain 3M™ Versaflo™ headtops.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité