3M™ Speedglas™ Heavy-Duty Back Pack
Fournisseur: 3M
BPK-HD
MMMABPK-HDEA 185 EUR InStock
MMMABPK-HD
3M™ Speedglas™ Heavy-Duty Back Pack
Matériel de soudage
Back pack harness optimised for welding environment.
  • Designed for comfort
  • Fits a wide variety of body sizes
  • Ventilated design against back
  • Carry/Hang Handle when stored in a dry and clean area
  • Made for heavy duty industries
  • Designed to be perceived as balanced when carried
  • Flame retardant harness designed for Welding environment withstanding spatter and sparks
  • Shoulder Harnesses adjustable position, length, width and a chest strap for extra tight fit well balanced on the back
Back pack designed for holding 3M™ Adflo™ and 3M™ Versaflo™ PAPR's.
The BPK-HD back pack , enables the PAPR to be worn on the BPK-HD backpack. This configuration allows the PAPR to be worn on the wearer's back rather than on a belt at the waist. The BPK-HD is optimised for Adflo and welding application, but may be used for selected additonal 3M PAPR e.g.
TR-series and heavy duty industry. This configuration may also enable the PAPR to be worn when a worker already wears other equipment around their waist. For customers in industries who need to have a heavy duty, optimised for welding PAPR backpack, the backpack delivers a robust, ergonomically designed, comfortable backpack that can fit a range of body sizes.
Attachment plate dimensions: 390×220 mm
Recommended application: Grinding, metal repair, welding
Recommended industry: Construction, heavy infrastructure, industrial maintenance, mining, oil and gas
Attention: For industrial/occupational use only. Not for consumer sale or use.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité